missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,3-Dimethyl-1-butene, 95%

Product Code. 11382656
Change view
Click to view available options
Quantity:
250mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382656 250mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11382656 Supplier Alfa Aesar™ Supplier No. L08693.AK

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Applications
It is used in the reaction pathways for the conversion of methanol and olefins on H-ZSM-5 zeolite.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Specifications

CAS 558-37-2
Boiling Point 40°C to 41°C
Flash Point −28°C (−18°F)
Molecular Formula C6H12
MDL Number MFCD00008853
Quantity 250mL
UN Number UN3295
Beilstein 1697101
Synonym 3,3-dimethyl-1-butene, neohexene, 1-butene, 3,3-dimethyl, tert-butylethylene, tert-butylethene, tert-hexene, trimethylvinylmethane, 3,3-dimethylbutene, 2,2-dimethyl-3-butene, unii-01zb73d2kk
InChI Key PKXHXOTZMFCXSH-UHFFFAOYSA-N
SMILES CC(C)(C)C=C
IUPAC Name 3,3-dimethylbut-1-ene
Molecular Weight (g/mol) 84.16
PubChem CID 11210
Formula Weight 84.16
Percent Purity 95%
Chemical Name or Material 3,3-Dimethyl-1-butene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.