missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butanol, 97%

Product Code. 15453887
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15453887 10 g 10g
15463887 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453887 Supplier Thermo Scientific Alfa Aesar Supplier No. H30194.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-95-3 | C6H14O | 102.18 g/mol

3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Dimethyl-1-butanol, is an organic building block used for the synthesis of various pharmaceutical compounds. It is an important intermediate in the synthesis of Neotame, an enhanced sweetening agent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-95-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002928
InChI Key DUXCSEISVMREAX-UHFFFAOYSA-N
Synonym 3,3-dimethyl-1-butanol, 3,3-dimethylbutanol, 1-butanol, 3,3-dimethyl, neohexanol, unii-543oyd666t, tert-hexyl alcohol, 3,3-dimethyl-butan-1-ol, chembl25029, tert-hexanol, pubchem3096
PubChem CID 12233
IUPAC Name 3,3-dimethylbutan-1-ol
SMILES CC(C)(C)CCO

Specifications

Melting Point -60°C
Density 0.81
Boiling Point 142°C to 143°C
Flash Point 56°C (133°F)
Odor Alcohol-like
Linear Formula (CH3)3CCH2CH2OH
Refractive Index 1.414
Quantity 10 g
UN Number UN1987
Solubility Information Slightly soluble in water.
Formula Weight 102.18
Percent Purity 97%
Chemical Name or Material 3,3-Dimethyl-1-butanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.