missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethoxybenzidine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30319933
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319933 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319933 Supplier Dr. Ehrenstorfer Supplier No. DREA12721000AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3'-Dimethoxybenzidine
CAS 119-90-4
Analyte or Component Names 3,3'-Dimethoxybenzidine
Shipping Condition Room Temperature
Molecular Formula C14 H16 N2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Benzidine, 3,3'-dimethoxy- (8CI), Fast Blue B Base (6CI), 3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diamine, 3,3'-Dimethoxy-4,4'-diaminobiphenyl, 3,3'-Dimethoxy-4,4'-diaminodiphenyl, 4,4'-Bi-o-anisidine, 4,4'-Diamino-3,3'-dimethoxy-1,1'-biphenyl, Amacel Developed Navy SD, Azogene Fast Blue B, Blue BN Base, Blue Base Irga B, Blue Base NB, C.I. Disperse Black 6, Cellitazol B, Cibacete Diazo Navy Blue 2B, Diacel Navy DC, Dianisidine, Fast Blue Base B, Fast Blue DSC Base, Hiltonil Fast Blue B Base, Kayaku Blue B Base, Lake Blue B Base, Mitsui Blue B Base, NSC 3168, Naphthanil Blue B Base, Setacyl Diazo Navy R, o-Dianisidine
InChI Formula InChI=1S/C14H16N2O2/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2/h3-8H,15-16H2,1-2H3
SMILES COc1cc(ccc1N)c2ccc(N)c(OC)c2
IUPAC Name 4-(4-amino-3-methoxyphenyl)-2-methoxyaniline
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 244.29
Quantity 1 mL
Formula Weight 244.1212 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.