missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluorocyclobutanecarboxylic acid, 97%, Thermo Scientific™

Product Code. 15266028 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15266028 250 mg 250mg
15256028 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15266028 Supplier Thermo Scientific Acros Supplier No. 452262500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 107496-54-8
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.1
InChI Key PLRCVBKYFLWAAT-UHFFFAOYSA-N
Synonym 3,3-difluorocyclobutanecarboxylic acid, 3,3-difluoro-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3,3-difluoro, cyclobutanecarboxylicacid, 3,3-difluoro, 3,3-difluorocyclobutanecarboxylicacid, acmc-209yhn, 3,3-difluorocyclopropyl methanol, 3,3-difluorocyclobutane carboxylic acid, 3,3-difluorocyclobutane-carboxylic acid, 3,3-difluoro-cyclobutane-1-carboxylic acid
PubChem CID 13777789
IUPAC Name 3,3-difluorocyclobutane-1-carboxylic acid
SMILES C1C(CC1(F)F)C(=O)O

Specifications

Melting Point 50.0°C to 52.5°C
Density 1.3700g/mL
Boiling Point 205.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Quantity 250 mg
Specific Gravity 1.37
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 3,3-Difluorocyclobutanecarboxylic acid
compliance-icons
Product Identifier
  • 3,3-Difluorocyclobutanecarboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.