missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoroazetidine Hydrochloride, TRC

Product Code. 30381146
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381146 100 mg
30380050 250 mg
30384985 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381146 Supplier Toronto Research Chemicals Supplier No. TRCD442330100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288315-03-7
Molecular Formula C3H5F2N . HCl
Molecular Weight (g/mol) 129.53
Synonym Azetidine, 3,3-difluoro-, hydrochloride (1:1) (ACI), Azetidine, 3,3-difluoro-, hydrochloride (9CI), 3,3-Difluoroazetidine hydrochloride
IUPAC Name 3,3-difluoroazetidine;hydrochloride
SMILES Cl.FC1(F)CNC1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C3H5F2N.ClH/c4-3(5)1-6-2-3;/h6H,1-2H2;1H
Formula Weight 129.0157
Chemical Name or Material 3,3-Difluoroazetidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.