missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoroazetidine hydrochloride, 95%

Product Code. 11451120
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451120 5 g 5g
11389959 250 mg 250mg
11399959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451120 Supplier Thermo Scientific Alfa Aesar Supplier No. H25751.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 288315-03-7 | C3H6ClF2N | 129.535 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288315-03-7
Molecular Formula C3H6ClF2N
MDL Number MFCD05663714
InChI Key CDBAEFXTCRKJPZ-UHFFFAOYSA-N
Synonym 3,3-difluoroazetidine hydrochloride, 3,3-difluoroazetidine hcl, azetidine, 3,3-difluoro-, hydrochloride, azetidine, 3,3-difluoro-, hydrochloride 1:1, 3,3-difluoroazetidine hydrochloride 1:1, 3,3-difluoroazetidinehydrochloride, acmc-1clfn, 3,3-difluoroazetidine-hcl, difluoroazetidine hydrochloride, 3,3-difluoroazetidin hydrochloride
PubChem CID 2758247
IUPAC Name 3,3-difluoroazetidine;hydrochloride

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
Sensitivity Hygroscopic
SMILES Cl.FC1(F)CNC1
Molecular Weight (g/mol) 129.53
Formula Weight 129.54
Percent Purity 95%
Chemical Name or Material 3,3-Difluoroazetidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.