missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylthiadicarbocyanine iodide

Product Code. 11370518
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11370518 2 g 2g
11360518 0.5 g 0.5g
11350518 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11370518 Supplier Thermo Scientific Alfa Aesar Supplier No. 043851.04
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

As a solvent soluble laser dye | CAS: 514-73-8 | C23H24IN2S2+ | 519.483 g/mol

It is employed as a solvent soluble laser dye. DTDCi is used as fluorescent contrast agent in three-dimensional fluorescence lifetime tomography and as a dye for rapid genetic screening. It is a potential sensitive probe useful for fluorometric analysis of transferable membrane pores and effects of membrane potential on Na cotransports in eel intestinal brush-border membrane vesicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 514-73-8
Molecular Formula C23H24IN2S2+
Molecular Weight (g/mol) 519.483
MDL Number MFCD00074829
InChI Key MNQDKWZEUULFPX-UHFFFAOYSA-N
Synonym 3,3'-diethylthiadicarbocyanine iodide
PubChem CID 91869351
IUPAC Name (2E)-3-ethyl-2-[(2E,4E)-5-(3-ethyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;hydroiodide
SMILES CCN1C2=CC=CC=C2SC1=CC=CC=CC3=[N+](C4=CC=CC=C4S3)CC.I

Specifications

Melting Point 248°C (decomposition)
Color Green
Quantity 2 g
UN Number UN2811
Beilstein 3838938
Merck Index 14,3369
Solubility Information Soluble in methanol
Formula Weight 518.47
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3,3'-Diethylthiadicarbocyanine iodide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.