missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethyloxadicarbocyanine iodide, 96%

Product Code. 15465877
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465877 250 mg 250mg
15445877 1 g 1g
15455877 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15465877 Supplier Thermo Scientific Alfa Aesar Supplier No. H33094.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14806-50-9 | C23H23IN2O2 | 486.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14806-50-9
Molecular Formula C23H23IN2O2
Molecular Weight (g/mol) 486.35
MDL Number MFCD00011953,MFCD00011953
InChI Key CLDZYSUDOQXJOU-UHFFFAOYSA-M
Synonym dodc iodide, 3-ethyl-2-5-3-ethyl-2-benzoxazolinylidene-1,3-pentadienyl benzoxazolium iodide, 5-3-ethylbenzoxazol-2-ylidene-1-3-ethylbenzoxazolium-2-yl-1,3-pentadiene iodide
PubChem CID 91972131
ChEBI CHEBI:51899
IUPAC Name (2Z)-3-ethyl-2-[5-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzoxazole;iodide
SMILES [I-].CCN1\C(OC2=CC=CC=C12)=C\C=C\C=CC1=[N+](CC)C2=CC=CC=C2O1

Specifications

Melting Point ∽215°C (decomposition)
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 486.36
Percent Purity 96%
Chemical Name or Material 3,3'-Diethyloxadicarbocyanine iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.