missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11416860
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416860 250 mg 250mg
11426860 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416860 Supplier Thermo Scientific Alfa Aesar Supplier No. L19579.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester is used as a synthetic and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester is used as a synthetic and pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153737-25-8
Molecular Formula C13H25BO4
Molecular Weight (g/mol) 256.149
MDL Number MFCD03788737
InChI Key ZXGPKUGXWIEUJA-MDZDMXLPSA-N
Synonym 3,3-diethoxy-1-propenylboronic acid pinacol ester, e-2-3,3-diethoxyprop-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,3-diethoxy-trans-1-propenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-3,3-diethoxy-1-propen-1-yl-4,4,5,5-tetramethyl, 2-e-3,3-diethoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-1e-3,3-diethoxyprop-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-3,3-diethoxy-1-propenylboronic acid pinacol ester, 3,3-diethoxy-1-propenylboronic acid picanol ester, e + z, 3-propene aldehyde-boronic acid pinacol ester diethyl acetal, e-2-3,3-diethoxyprop-1-enyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11065022
IUPAC Name 2-[(E)-3,3-diethoxyprop-1-enyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CC(OCC)OCC

Specifications

Refractive Index 1.441
Quantity 250 mg
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 256.15
Percent Purity 97%
Chemical Name or Material 3,3-Diethoxy-trans-1-propenylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.