missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine tetrahydrochloride hydrate, 97%

Product Code. 15454629
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 15454629

Brand: Thermo Scientific Alfa Aesar J62216.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,3'-Diaminobenzidine tetrahydrochloride hydrate, CAS # 868272-85-9, is a peroxidase substrate that is commonly used for immunoblotting and immunohistochemical staining techniques. | CAS: 868272-85-9 | C12H18Cl4N4 | 360.10 g/mol

3,3?-diaminobenzidine-tetrahydrochloride-dihydrate (DAB) was used as a chromogen, for detecting the antigen-antibody complex. It is one of the most potent marker being used in immunochemistry, mostly at the electron and light microscope levels. It is used as a histological stain for peroxidase activity. Reaction between 3,3'-Diaminobenzidine and peroxidase forms a visible insoluble product. This approach has been used to assay activity of glutamate decarboxylase in rat cerebellar sections and to detect horseradish peroxidase presence on cultured cell surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3,3'-Diaminobenzidine tetrahydrochloride hydrate is an organic compound used as a peroxidase substrate

Application

• 3,3'-Diaminobenzidine tetrahydrochloride hydrate is commonly used for immunoblotting and immunohistochemical staining techniques

• It has also been used for the histological stain to measure the peroxidase activity. The interaction between 3,3'-Diaminobenzidine tetrahydrochloride hydrate and peroxidase leads to the formation of a visible insoluble product. Therefore, it has been applied to measure glutamate decarboxylase activity in rat cerebellar sections and to detect horseradish peroxidase presence on cultured cell surfaces

• It is used as a chromogen for detecting the antigen-antibody complex

• This compound is also used in several laboratory applications, including electron and light microscope, and FISH (fluorescence in situ hybridization)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868272-85-9
Molecular Formula C12H18Cl4N4
Molecular Weight (g/mol) 360.10
MDL Number MFCD08273058
InChI Key KJDSORYAHBAGPP-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine tetrahydrochloride hydrate, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride hydrate, acmc-209qbm, 3,3'-diaminobenzidine hydrate tetrahydrochloride, biphenyl-3,3',4,4'-tetramine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, isopac r, 1,1'-biphenyl-3,3',4,4'-tetraamine tetrahydrochloride xhydrate, 3,3 inverted exclamation marka-diaminobenzidine tetrahydrochloride hydrate, 3,3'-diaminobenzidine tetrahydrochloride hydrate, for spectrophotometric det. of se
PubChem CID 23191111
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine;hydrate
SMILES Cl.Cl.Cl.Cl.NC1=C(N)C=C(C=C1)C1=CC(N)=C(N)C=C1

Specifications

Color Gray
Quantity 10 g
Solubility Information Soluble at 5%(w/v) in water
Formula Weight 360.11 (Anhydrous)
Physical Form Powder
Chemical Name or Material 3,3'-Diaminobenzidine tetrahydrochloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.