missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine, tablets, Thermo Scientific™

Product Code. 15423649
Change view
Click to view available options
Quantity:
100 Ea.
50 Ea.
Unit Size:
Pack of 100
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423649 50 Ea. Pack of 50
15433649 100 Ea. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423649 Supplier Thermo Scientific Alfa Aesar Supplier No. J60972.KW
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,3′-Diaminobenzidine is used to detect fingerprints in blood. 3,3′-Diaminobenzidine is used in immunohistology and immunoblotting as a precipitating substrate for the localization of peroxidase activity. Slides stained with DAB may be coverslipped in the traditional manner and stored for future reference. Many enhancement methods have been reported for DAB involving different buffer conditions, the addition of metal ions and post-treatment applications. DAB has been used in immunohistochemical staining of nucleic acids and proteins. Application of the silver-gold intensified 3,3′ -diaminobenzidine chromogen to the light and electron microscopic detection of the luteinizing hormone-releasing hormone system of the rat brain. 3, 3′-diaminobenzidine (DAB)-H2O2-horseradish peroxidase (HRP) has been presented and used for the sensitive detection of carcinoembryonic antigen (CEA) in human serum.

Notes
Each tablet contains 5mg of DAB, each tablet weighs 240mg. One tablet will yield 10 ml of substrate solution, enough to effectively detect 50cm² of membrane or about 20 slides. Insoluble product. Reaction Condition: Dissolve one 5mg tablet in 10 ml of 50mM Tris, 0.1M NaCl, pH 7.6 and 0.01% hydrogen peroxide. The solution may be filtered through a 0.2µm filter immediately prior to use if necessary. The presence of nickel or cobalt increases sensitivity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-95-2
Molecular Formula C12H14N4
Molecular Weight (g/mol) 214.272
MDL Number MFCD00007725
InChI Key HSTOKWSFWGCZMH-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
PubChem CID 7071
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N

Specifications

Color Purple
Quantity 50 Ea.
Beilstein 1212988
Sensitivity Light sensitive
Solubility Information Soluble in water (partly).
Formula Weight 214.27
Physical Form Tablets
Chemical Name or Material 3,3'-Diaminobenzidine, tablets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.