missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine, 98+%

Product Code. 15420438
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420438 100 g 100g
15400438 5 g 5g
15410438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420438 Supplier Thermo Scientific Alfa Aesar Supplier No. H54000.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-95-2 | C12H14N4 | 214.272 g/mol

3,3'-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

Solubility
Soluble in water (partly).

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-95-2
Molecular Formula C12H14N4
Molecular Weight (g/mol) 214.272
MDL Number MFCD00007725
InChI Key HSTOKWSFWGCZMH-UHFFFAOYSA-N
Synonym 3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
PubChem CID 7071
IUPAC Name 4-(3,4-diaminophenyl)benzene-1,2-diamine
SMILES C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N

Specifications

Melting Point 175°C to 177°C
Quantity 100 g
Sensitivity Light sensitive
Solubility Information Soluble in water (partly).
Formula Weight 214.27
Percent Purity 99%
Chemical Name or Material 3,3'-Diaminobenzidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.