missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-n-propylthiacarbocyanine iodide, 96%, Thermo Scientific Chemicals

Product Code. p-7031353
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15425127

Brand: Thermo Scientific Alfa Aesar H32400.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53336-12-2
Molecular Formula C23H25IN2S2
Molecular Weight (g/mol) 520.49
MDL Number MFCD00013340
InChI Key JGTCEHAVVINOPG-UHFFFAOYSA-M
Synonym 3,3'-dipropylthiacarbocyanine iodide, 3-propyl-2-3-3-propyl-2 3h benzothiazolylidene-1-propenyl benzothiazolium iodide, 3-propyl-2-1e,3z-3-3-propylbenzo d thiazol-2 3h-ylidene prop-1-en-1-yl benzo d thiazol-3-ium iodide
PubChem CID 57369736
SMILES [I-].CCCN1C(SC2=CC=CC=C12)=CC=CC1=[N+](CCC)C2=CC=CC=C2S1

Specifications

Quantity 250 mg
IUPAC Name 3-propyl-2-[3-(3-propyl-1,3-benzothiazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzothiazole;iodide
Formula Weight 520.5
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-propylthiacarbocyanine iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.