missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Di-n-pentylthiadicarbocyanine iodide, 96%, Thermo Scientific™

Product Code. 15438725
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438725 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15438725 Supplier Thermo Scientific Alfa Aesar Supplier No. H33978.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 53213-96-0
Melting Point ∽212°C (decomposition)
Molecular Formula C29H35IN2S2
MDL Number MFCD07433877
Quantity 500 mg
Sensitivity Light sensitive
Synonym 3-pentyl-2-1e,3e-5-3-pentyl-1,3-benzothiazol-2 3h-ylidene-1,3-pentadienyl-1,3-benzothiazol-3-ium iodide, 3-pentyl-2-5-3-pentyl-1,3-benzothiazol-3-ium-2-yl penta-2,, 3-pentyl-2-1e,3e,5z-5-3-pentylbenzo d thiazol-2 3h-ylidene penta-1,3-dien-1-yl benzo d thiazol-3-ium iodide, 3-pentyl-2-5-3-pentyl-1,3-benzothiazol-2 3h-ylidene penta-1,3-dien-1-yl-1,3-benzothiazol-3-ium iodide
InChI Key IDGJDXHDJJPLIE-UHFFFAOYSA-M
SMILES [I-].CCCCCN1C(SC2=CC=CC=C12)=CC=CC=CC1=[N+](CCCCC)C2=CC=CC=C2S1
IUPAC Name 3-pentyl-2-[5-(3-pentyl-1,3-benzothiazol-3-ium-2-yl)penta-2,4-dienylidene]-1,3-benzothiazole;iodide
Molecular Weight (g/mol) 602.64
PubChem CID 53425757
Formula Weight 602.65
Percent Purity 96%
Chemical Name or Material 3,3'-Di-n-pentylthiadicarbocyanine iodide
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.