missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid, 97%, Thermo Scientific™

Product Code. 15440586
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440586 250 mg 250mg
15430586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440586 Supplier Thermo Scientific Alfa Aesar Supplier No. H53320.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957060-87-6
Molecular Formula C13H14B2N2O5
Molecular Weight (g/mol) 299.88
MDL Number MFCD09027259
InChI Key UQUQNYFFKZOTSS-UHFFFAOYSA-N
Synonym 1,3-bis 3-boronophenyl urea, carbonylbis azanediyl bis 3,1-phenylene diboronic acid, 3-3-dihydroxyboranyl phenyl carbamoyl amino phenylboronic acid, carbonylbis azanediyl-3,1-phenylene diboronic acid
PubChem CID 44119184
IUPAC Name [3-[(3-boronophenyl)carbamoylamino]phenyl]boronic acid
SMILES OB(O)C1=CC(NC(=O)NC2=CC=CC(=C2)B(O)O)=CC=C1

Specifications

Melting Point 225°C to 226°C
Quantity 250 mg
Formula Weight 299.88
Percent Purity 97%
Chemical Name or Material 3,3'-Carbonylbis(azanediyl)bis(3,1-phenylene)diboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.