missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)-5-methyl-1,2,4-oxadiazole, ≥97%, Thermo Scientific™

Product Code. 11300617
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11300617 250 mg 250mg
10708655 1 g 1g
10763327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11300617 Supplier Thermo Scientific Maybridge Supplier No. CC39110CB
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 160377-57-1
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD08087563
InChI Key GTYLSVIVKRJHIQ-UHFFFAOYSA-N
Synonym 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-3-bromophenyl-5-methyl, acmc-1c5gb, chembrdg-bb 9035421, 3-3-bromophenyl-5-methyl-1,2,4-oxadiazol, 3-3-bromophenyl-5-methyl-1,2,4 oxadiazole, 3-3-bromophenyl-5-methyl-1,2,4-oxadiazole saltdata: free
PubChem CID 8895139
IUPAC Name 3-(3-bromophenyl)-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC(=CC=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 239.07
Percent Purity 97%
Chemical Name or Material 3-(3-Bromophenyl)-5-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.