missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10062314
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 425379-68-6
Molecular Formula C10H9BrN2
Molecular Weight (g/mol) 237.10
MDL Number MFCD09065008
InChI Key PNVHBUUFMMMKBR-UHFFFAOYSA-N
Synonym 3-3-bromophenyl-1-methyl-1h-pyrazole, 3-3-bromophenyl-1-methylpyrazole, 1h-pyrazole,3-3-bromophenyl-1-methyl
PubChem CID 22329555
IUPAC Name 3-(3-bromophenyl)-1-methylpyrazole
SMILES CN1C=CC(=N1)C1=CC(Br)=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.1
Percent Purity 97%
Chemical Name or Material 3-(3-Bromophenyl)-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'