missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenoxy)-N,N-dimethylpropylamine, 97%, Thermo Scientific™

Product Code. 10402844
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10402844 5 g Amber Glass Bottle 5g
10728845 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402844 Supplier Thermo Scientific Maybridge Supplier No. CC59410DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-57-4
Molecular Formula C11H16BrNO
Molecular Weight (g/mol) 258.159
MDL Number MFCD03718729
InChI Key FMYWALTVPROHIA-UHFFFAOYSA-N
Synonym 3-3-bromophenoxy-n,n-dimethylpropylamine, 3-3-bromophenoxy-n,n-dimethylpropan-1-amine, 3-3-bromophenoxy propyl dimethylamine
PubChem CID 2183561
IUPAC Name 3-(3-bromophenoxy)-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCOC1=CC(=CC=C1)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 258.16
Percent Purity 97%
Chemical Name or Material 3-(3-Bromophenoxy)-N, N-dimethylpropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.