missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Product Code. 10299372
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299372 250 mg Amber Glass Bottle 250mg
10700374 1 g Amber Glass Bottle 1g
11326835 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299372 Supplier Thermo Scientific Maybridge Supplier No. CC39108CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 253273-90-4
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
InChI Key CERZNQPNTHWEAD-UHFFFAOYSA-N
Synonym 3-3-bromomethyl phenyl-5-methyl-1,2,4-oxadiazole, 3-3-bromomethylphenyl-5-methyl-1,2,4-oxadiazole, 3-5-methyl-1,2,4-oxadiazol-3-yl benzyl bromide, 1,2,4-oxadiazole,3-3-bromomethyl phenyl-5-methyl, 3-3-bromomethyl phenyl-5-methyl-1,2,4-oxadiazol
PubChem CID 7060548
IUPAC Name 3-[3-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC(=CC=C2)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 3-[3-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.