Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 3-Way Stopcocks
Product Code. p-8005949
Change view
Click to view available options
Plug Bore:
2.5 mm
4 mm
Includes:
PTFE Plug, Screw Thread Retaining Device
Solid Glass Plug, Screw Thread Retaining Device
Solid Glass Plug, Screw Thread Rubber Retaining Device
Unit Size:
Each
Product Code. Plug Bore Includes unitSize
10777851 2.5 mm Solid Glass Plug, Screw Thread Retaining Device Each
10088170 4 mm PTFE Plug, Screw Thread Retaining Device Each
10509971 2.5 mm PTFE Plug, Screw Thread Retaining Device Each
10612472 4 mm Solid Glass Plug, Screw Thread Retaining Device Each
10597911 4 mm Solid Glass Plug, Screw Thread Rubber Retaining Device Each
5 options
Product Code. 10777851
Brand: Lenz Laborglasinstrumente™ 02501402
Only null left
Add to basket
Add to basket
Specifications
Type 3-way Stopcock
Plug Bore 2.5 mm
Material Borosilicate Glass
Includes Solid Glass Plug, Screw Thread Retaining Device