missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Benzothienyl)-N-Fmoc-L-alanine, 95%

Product Code. p-7034371
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15488547 250 mg 250mg
15468547 1 g 1g
15478547 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15488547

Brand: Thermo Scientific Alfa Aesar H52054.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 177966-60-8 | C26H21NO4S | 443.52 g/mol

It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility (second only to glycine) when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility (second only to glycine) when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177966-60-8
Molecular Formula C26H21NO4S
Molecular Weight (g/mol) 443.52
MDL Number MFCD00672562
InChI Key BQIZNDWONIMCGM-QHCPKHFHSA-N
Synonym fmoc-l-3-benzothienylalanine, fmoc-3-ala 3-benzothienyl-oh, fmoc-l-3-3-benzothienyl alanine, fmoc-beta-3-benzothienyl-l-alanine, fmoc-beta-3-benzothienyl-ala-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzo b thiophen-3-yl propanoic acid, 2s-3-1-benzothiophen-3-yl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-3-benzothienylala, fmoc-l3-benzothienylalanine, fmoc-3-3-benzothienyl-l-alanine
PubChem CID 2761478
IUPAC Name (2S)-3-(1-benzothiophen-3-yl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES OC(=O)[C@H](CC1=CSC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
Beilstein 9296209
Formula Weight 443.52
Percent Purity 95%
Chemical Name or Material 3-(3-Benzothienyl)-N-Fmoc-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.