missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Aminopropoxy)-N-[2-[[3-[[(4-chloro-2-hydroxyphenyl)amino]carbonyl]-2-naphthalenyl]oxy]ethyl]-2-naphthalenecarboxamide Hydrochloride, TRC

Product Code. 30316114
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30316114

Brand: Toronto Research Chemicals TRCA6164435MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Aminopropoxy)-N-[2-[(3-[((4-chloro-2-hydroxyphenyl)amino)carbonyl]-2-naphthalenyl)oxy]ethyl]-2-naphthalenecarboxamide Hydrochloride
CAS 1433286-70-4
Molecular Formula C33H30ClN3O5 . HCl
Quantity 5 mg
Synonym 666-15
InChI Formula InChI=1S/C33H30ClN3O5.ClH/c34-25-10-11-28(29(38)20-25)37-33(40)27-17-22-7-2-4-9-24(22)19-31(27)42-15-13-36-32(39)26-16-21-6-1-3-8-23(21)18-30(26)41-14-5-12-35;/h1-4,6-11,16-20,38H,5,12-15,35H2,(H,36,39)(H,37,40);1H
SMILES Cl.NCCCOc1cc2ccccc2cc1C(=O)NCCOc3cc4ccccc4cc3C(=O)Nc5ccc(Cl)cc5O
IUPAC Name 3-(3-aminopropoxy)-N-[2-[3-[(4-chloro-2-hydroxyphenyl)carbamoyl]naphthalen-2-yl]oxyethyl]naphthalene-2-carboxamide;hydrochloride
Molecular Weight (g/mol) 620.5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.