missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Mikromol™
Description
3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride |
| CAS | 115189-28-1 |
| Analyte or Component Names | 3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 Cl2 N2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Clomipramine Hydrochloride Imp. D (EP), Clomipramine Imp. D (EP), Clomipramine USP Related Compound D, Clomipramine USP RC D, 3-(3,7-Dichloro-10,11-dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Clomipramine Hydrochloride Imp. D (EP) as Hydrochloride, 5H-Dibenz[b,f]azepine-5-propanamine, 3,7-dichloro-10,11-dihydro-N,N-dimethyl-, hydrochloride (1:1), 5H-Dibenz[b,f]azepine, 3,7-dichloro-5-(3-dimethylaminopropyl)-10,11-dihydro-, hydrochloride (6CI), Clomipramine Related Compound D as Hydrochloride, Clomipramine Hydrochloride Impurity D as Hydrochloride, Clomipramine Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C19H22Cl2N2.ClH/c1-22(2)10-3-11-23-18-12-16(20)8-6-14(18)4-5-15-7-9-17(21)13-19(15)23;/h6-9,12-13H,3-5,10-11H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCCN1c2cc(Cl)ccc2CCc3ccc(Cl)cc13 |
| IUPAC Name | 3-(2,9-dichloro-5,6-dihydrobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?