missing translation for 'onlineSavingsMsg'
Learn More

3,3’,5-Triiodo Thyropropionic Acid, TRC

Product Code. 30041294
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041294 5 mg 5mg
18031230 50 mg 50mg
18032520 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041294 Supplier Toronto Research Chemicals Supplier No. TRCT7954105MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-26-3
Molecular Formula C15 H11 I3 O4
Molecular Weight (g/mol) 635.96
IUPAC Name 3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES OC(=O)CCc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1

Specifications

Quantity 5 mg
Synonym Hydrocinnamic acid, 4-(4-hydroxy-3-iodophenoxy)-3,5-diiodo- (6CI,8CI), 4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodobenzenepropanoic acid, 3,3',5-Triiodothyropropionic acid, 4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodohydrocinnamic acid, Atralose, Birodan, Thyropropic acid, Triopron, Triprop, [(4-Hydroxy-3-iodo-4-phenoxy)-3,5-diiodophenyl]propionic acid, β-[4-(3'-Iodo-4'-hydroxyphenoxy)-3,5-diiodophenyl]propionic acid, 3,3',5-Triiodothyropropionic Acid, 4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodobenzenepropanoic acid
InChI Formula InChI=1S/C15H11I3O4/c16-10-7-9(2-3-13(10)19)22-15-11(17)5-8(6-12(15)18)1-4-14(20)21/h2-3,5-7,19H,1,4H2,(H,20,21)
Formula Weight 635.7791
Percent Purity >95%
Chemical Name or Material 3,3',5-Triiodothyropropionic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.