missing translation for 'onlineSavingsMsg'
Learn More

3,3',5-Triiodo-L-thyronine, TRC

Product Code. 30010667
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010667 100 mg 100mg
30004472 250 mg 250mg
30009719 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010667 Supplier Toronto Research Chemicals Supplier No. TRCT795380100MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6893-02-3
Molecular Formula C15 H12 I3 N O4
Molecular Weight (g/mol) 650.97
Synonym Liothyronine, L-Tyrosine, O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-, Alanine, 3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-, L- (8CI), Thyronine, 3,3',5-triiodo-, L- (6CI), O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, 3,3',5-Triiodo-L-thyronine, 3,3',5-Triiodothyronine, 3,5,3'-Triiodothyronine, 4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenylalanine, Cyronine, L-3,3',5-Triiodothyronine, L-Liothyronine, L-T3, L-Triiodothyronine, Liothyronin, NSC 80203, T3, T3 (Hormone), T3 (amino acid), Tresitope, Triiodo-L-thyronine, Triiodothyronine, Tri-iodothyronine
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoic acid
SMILES N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(=O)O

Specifications

InChI Formula InChI=1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1
Quantity 100 mg
Formula Weight 650.79
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material 3,3',5-Triiodo-L-thyronine (T3)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.