missing translation for 'onlineSavingsMsg'
Learn More

3,3’,5-Triiodo-L-thyronine Sodium Salt, TRC

Product Code. 30133009
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133009 1 g 1g
17953375 500 mg 500mg
17973455 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133009 Supplier Toronto Research Chemicals Supplier No. TRCT7953851G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-06-1
Molecular Formula C15 H11 I3 N O4 . Na
Molecular Weight (g/mol) 672.96
Synonym Liothyronine Sodium, L-Tyrosine, O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-, sodium salt (1:1), Alanine, 3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-, monosodium salt, L- (8CI), L-Tyrosine, O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-, monosodium salt (9CI), O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt (1:1), 3-[4-(4-Hydroxy-3-iodophenoxy)-3,5-diiodophenyl]-L-alanine monosodium salt, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine monosodium salt), 3,3',5-Triiodo-L-thyronine sodium salt, 3,3',5-Triiodothyronine sodium salt, 3,5,3'-Triiodo-L-thyronine monosodium salt, Basoprocin, Cynomel, Cyomel, Cytobin, Cytomel, Cytomel sodium, Cytomine, Ibiothyron, L-Triiodothyronine sodium salt, Liothyroniinde sodium salt, Liothyronin sodium, Liothyronine sodium, Sodium 3,3',5-triiodo-L-thyronine, Sodium 3,5,3'-triiodothyronine, Sodium L-3,3',5-triiodothyronine, Sodium L-triiodothyronine, Sodium T3, Sodium liothyronine, Sodium triiodo-L-thyronine, Sodium triiodothyronine, T3 Sodium salt, Tertroxin, Triiodothyronine sodium salt, Triostat, Triothyrone
IUPAC Name sodium;(2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
SMILES [Na+].N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(=O)[O-]

Specifications

InChI Formula InChI=1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1
Quantity 1 g
Formula Weight 672.772
Shipping Condition Room Temperature
Chemical Name or Material Liothyronine sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.