missing translation for 'onlineSavingsMsg'
Learn More

3-(3,5-Difluorophenyl)propionic acid, 97%, Thermo Scientific™

Product Code. 15470438
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470438 5 g 5g
15480438 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470438 Supplier Thermo Scientific Alfa Aesar Supplier No. H54004.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(3,5-Difluorophenyl)propionic acid is used as intermediate in pharmaceuticals.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84315-24-2
Molecular Formula C9H8F2O2
Molecular Weight (g/mol) 186.158
MDL Number MFCD04116054
InChI Key SAAKANGUQMVTHQ-UHFFFAOYSA-N
PubChem CID 3669498
IUPAC Name 3-(3,5-difluorophenyl)propanoic acid
SMILES C1=C(C=C(C=C1F)F)CCC(=O)O

Specifications

Melting Point 60°C
Quantity 5 g
Synonym 3-3,5-difluorophenyl propionic acid, 3-3,5-difluorophenyl propanoic acid, benzenepropanoic acid, 3,5-difluoro, 3-3,5-difluoro-phenyl-propionic acid, 3-3,5-difluorophenyl propionicacid, acmc-209vrt, 3,5-difluorohydrocinnamic acid, 3,5-difluoro-benzenepropanoic acid, benzenepropanoic acid,3,5-difluoro, 3-3,5-difluoro-phenyl propionic acid
Formula Weight 186.16
Percent Purity 97%
Chemical Name or Material 3-(3,5-Difluorophenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.