missing translation for 'onlineSavingsMsg'
Learn More

3,3,5,5-Tetramethylcyclohexanone, 98%

Product Code. 10370904 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10370904 50 mL Glass bottle 50mL
10636532 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10370904 Supplier Thermo Scientific Acros Supplier No. 138390500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14376-79-5 | C10H18O | 154.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14376-79-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001640
InChI Key OQJMHUOCLRCSED-UHFFFAOYSA-N
Synonym 3,3,5,5-tetramethylcyclohexanone, cyclohexanone, 3,3,5,5-tetramethyl, 3,3,5,5-tetramethyl cyclohexanone, cyclohexanone,3,3,5,5-tetramethyl, acmc-1c4rg, 3,5,5-tetramethylcyclohexanone, cyclohexanone,3,5,5-tetramethyl, 3,3,5,5-tetramethyl-cyclohexanone, 3,3,5,5-tetramethyl-1-cyclohexanone, 3,3,5,5-tetramethyl-cyclohexan-1-one
PubChem CID 84399
IUPAC Name 3,3,5,5-tetramethylcyclohexan-1-one
SMILES CC1(CC(=O)CC(C1)(C)C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11°C to 12°C
Color Colorless to Yellow
Density 0.8954g/mL
Boiling Point 200°C to 203°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)4C6H6(=O)
Refractive Index 1.4510 to 1.453
Quantity 50 mL
Beilstein 07, IV, 89
Specific Gravity 0.8954
Formula Weight 154.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3,3,5,5-Tetramethylcyclohexanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.