missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbiphenyl, 97+%

Product Code. 11397256
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397256 1 g 1g
11307266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397256 Supplier Thermo Scientific Alfa Aesar Supplier No. L09172.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25570-02-9 | C16H18 | 210.32 g/mol

3,3',5,5'-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3',5,5'-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3',5,5'-tetracarboxylic acid by the oxidation of with potassium permanganate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′,5,5′-Tetramethylbiphenyl is used as a precursor for the preparation of 3,3′,5,5′-tetramethylenebiphenyl tetraanion. It is also used to prepare biphenyl-3,3′,5,5′-tetracarboxylic acid by the oxidation of with potassium permanganate.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25570-02-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00051910
InChI Key CMZYGFLOKOQMKF-UHFFFAOYSA-N
PubChem CID 520212
IUPAC Name 1-(3,5-dimethylphenyl)-3,5-dimethylbenzene
SMILES CC1=CC(=CC(C)=C1)C1=CC(C)=CC(C)=C1

Specifications

Melting Point 47°C to 49°C
Quantity 1 g
Beilstein 1935931
Synonym 3,3',5,5'-tetramethylbiphenyl, 1-3,5-dimethylphenyl-3,5-dimethylbenzene, 3,5,3',5'-tetramethylbiphenyl, 1,1'-biphenyl, 3,3',5,5'-tetramethyl, biphenyl, 3,3',5,5'-tetramethyl, acmc-1cihy, 2,2'5.5'-tetramethylbiphenyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl, 1,1'-biphenyl,3,3',5,5'-tetramethyl, 3,3',5,5'-tetramethyl-1,1'-biphenyl #
Formula Weight 210.32
Percent Purity ≥97%
Chemical Name or Material 3,3',5,5'-Tetramethylbiphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.