missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetramethylbenzidine, 98%

Product Code. p-7021509
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11431548

Brand: Thermo Scientific Alfa Aesar A13868.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,3',5,5'-Tetramethylbenzidine, 98%, or TMB, CAS # 54827-17-7, is a colorimetric reagent used in laboratory-based substrate detection. | CAS: 54827-17-7 | C16H20N2 | 240.35 g/mol

Excellent colorimetric substrate for detection of horseradish peroxidase labelled probes; particularly useful in ELISA techniques, blue result can be read spectrophotometrically at 370 or 650 nm3,3',5,5'-Tetramethylbenzidine is used as a clinical reagent for testing blood. It is also a chromogenic substrate which is used in staining procedures in immunohistochemistry. Further, it is used as a visualizing reagent in enzyme-linked immunosorbent assays. In addition to this, it is used as a replacement for carcinogenic compounds such as benzidine and o-phenylenediamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 3,3',5,5'-Tetramethylbenzidine, also known as 4,4'-Diamino-3,3',5,5'-tetramethylbiphenyl or TMB, is a colorimetric substrate for detection of horseradish peroxidase-labeled probes
  • Oxidation of 3,3',5,5'-Tetramethylbenzidine results in a pale blue end product that can be detected at 370 or 655 nm. Sulfuric acid (H2SO4) is used to stop the oxidation reaction and stabilize the reading, resulting in a yellow color that can be read at 450 nm

Application

  • 3,3',5,5'-Tetramethylbenzidine is used as a chromogenic visualization agent in enzyme-linked immunosorbent assays (ELISA) or immunohistochemistry (IHC) techniques
  • 3,3',5,5'-Tetramethylbenzidine has been used to detect molecules in aqueous samples such as copper ion and hypochlorite
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54827-17-7
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00007748
InChI Key UAIUNKRWKOVEES-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethylbenzidine, 3,3',5,5'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 3,5,3',5'-tetramethylbenzidine, tmb, 3,3',5,5'-tetramethyl benzidine, tmb substrate, bm blue, ccris 4727, tmb plus, tmb, insoluble
PubChem CID 41206
IUPAC Name 4-(4-amino-3,5-dimethylphenyl)-2,6-dimethylaniline
SMILES CC1=CC(=CC(=C1N)C)C2=CC(=C(C(=C2)C)N)C

Specifications

Melting Point 168°C to 171°C
Quantity 1 g
Beilstein 2808541
Sensitivity Light and moisture sensitive
Solubility Information Soluble in ethyl acetate,ethanol,acetone,chloroform,toluene and dimethyl sulfoxide. Insoluble in water.
Formula Weight 240.35
Percent Purity 98%
Chemical Name or Material 3,3',5,5'-Tetramethylbenzidine
compliance-icons compliance-icons
Product Identifier
  • 3,3',5,5'-Tetramethylbenzidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : 3,3',5,5'-tetramethyl-[1,1'-Biphenyl]-4,4'-diamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.