missing translation for 'onlineSavingsMsg'
Learn More

3,3',5,5'-Tetrabromobisphenol A, 97%

Product Code. 11403647
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403647 100 g 100g
11413647 500 g 500g
11423647 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403647 Supplier Thermo Scientific Alfa Aesar Supplier No. A17395.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-94-7 | C15H12Br4O2 | 543.875 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-94-7
Molecular Formula C15H12Br4O2
Molecular Weight (g/mol) 543.875
MDL Number MFCD00013962
InChI Key VEORPZCZECFIRK-UHFFFAOYSA-N
Synonym tetrabromobisphenol a, 3,3',5,5'-tetrabromobisphenol a, bromdian, 4,4'-propane-2,2-diyl bis 2,6-dibromophenol, 4,4'-isopropylidenebis 2,6-dibromophenol, firemaster bp 4a, tetrabromodian, 2,2-bis 3,5-dibromo-4-hydroxyphenyl propane, fire guard 2000, great lakes ba-59p
PubChem CID 6618
ChEBI CHEBI:33217
IUPAC Name 2,6-dibromo-4-[2-(3,5-dibromo-4-hydroxyphenyl)propan-2-yl]phenol
SMILES CC(C)(C1=CC(=C(C(=C1)Br)O)Br)C2=CC(=C(C(=C2)Br)O)Br

Specifications

Melting Point 179°C to 182°C
Density 1.76
Odor Odorless
Quantity 100 g
UN Number UN3077
Formula Weight 543.87
Percent Purity 97%
Chemical Name or Material 3,3',5,5'-Tetrabromobisphenol A
compliance-icons compliance-icons
Product Identifier
  • 3,3',5,5'-Tetrabromobisphenol A
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: TetrabromobisphenolA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.