missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3,4-Dimethoxyphenyl)propionic acid, 99%, Thermo Scientific™
Click to view available options
50g; Glass bottle
Specifications
CAS | 2107-70-2 |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C11H14O4 |
Linear Formula | (CH3O)2C6H3CH2CH2CO2H |
MDL Number | MFCD00002774 |
Quantity | 50g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'