missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dihydroxyphenyl)propionic acid, 98+%

Product Code. 11444743
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444743 5 g 5g
11454743 25 g 25g
11464743 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444743 Supplier Thermo Scientific Alfa Aesar Supplier No. A11638.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1078-61-1 | C9H10O4 | 182.175 g/mol

3-(3,4-Dihydroxyphenyl)propionic acid it can be used to produce 3-(3,4-dihydroxy-phenyl)-propionic acid octyl ester by heating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3,4-Dihydroxyphenyl)propionic acid it can be used to produce 3-(3,4-dihydroxy-phenyl)-propionic acid octyl ester by heating.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1078-61-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002776
InChI Key DZAUWHJDUNRCTF-UHFFFAOYSA-N
Synonym dihydrocaffeic acid, 3-3,4-dihydroxyphenyl propionic acid, hydrocaffeic acid, 3,4-dihydroxyhydrocinnamic acid, 3-3,4-dihydroxyphenyl propanoic acid, benzenepropanoic acid, 3,4-dihydroxy, hydrocaffeic acid polymer, hykop, 3,4-dihydroxyhydrocinnamate, 3,4-dihydroxybenzenepropionic acid
PubChem CID 348154
ChEBI CHEBI:48400
IUPAC Name 3-(3,4-dihydroxyphenyl)propanoic acid
SMILES C1=CC(=C(C=C1CCC(=O)O)O)O

Specifications

Melting Point 136°C to 140°C
Quantity 5 g
Beilstein 2213449
Solubility Information Slightly soluble in water.
Formula Weight 182.18
Percent Purity ≥98%
Chemical Name or Material 3- (3,4-Dihydroxyphenyl)propionic acid
compliance-icons
Product Identifier
  • 3-(3,4-Dihydroxyphenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3-(3,4-Dihydroxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.