missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dihydroxyphenyl)-L-alanine, 99%

Product Code. 10703724 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10703724 100 g Glass Bottle 100g
10606762 5 g Glass Bottle 5g
10701251 25 g Glass Bottle 25g
10485555 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10703724 Supplier Thermo Scientific Acros Supplier No. 167531000
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

3-(3,4-Dihydroxyphenyl)-L-alanine (levodopa orL-dopa), CAS # 59-92-7, is a dopamine precursor molecule. | CAS: 59-92-7 | C9H11NO4 | 197.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body

• Upon conversion to dopamine, it acts as an excitatory neurotransmitter

Applications

• L-dopa is useful in the laboratory study of Parkinson’s to replenish dopamine in depleted circuits

• L-dopa has also been used for laboratory research on hypertension and melanocyte activity

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-92-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00002598
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
Synonym levodopa, l-dopa, dopar, 3-hydroxy-l-tyrosine, 3,4-dihydroxy-l-phenylalanine, larodopa, bendopa, levopa, dihydroxy-l-phenylalanine, cidandopa
PubChem CID 6047
ChEBI CHEBI:15765
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Color Cream to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Loss on Drying 1% max.
Packaging Glass Bottle
Linear Formula (HO)2C6H3CH2CH(NH2)CO2H
Beilstein 14, IV, 2492
Merck Index 15, 5516
Sulfated Ash 0.1% max.
Quantity 100 g
Formula Weight 197.19
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3-(3, 4-Dihydroxyphenyl)-L-alanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.