missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4-Dichlorophenyl)triaz-2-ene-1-carbonitrile Potassium Salt, TRC

Product Code. 30381396
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381396 500 mg 500mg
30377006 1 g 1g
30381069 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381396 Supplier Toronto Research Chemicals Supplier No. TRCD482136500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3,4-Dichlorophenyl)triaz-2-ene-1-carbonitrile Potassium Salt
Analyte or Component Names 3-(3,4-Dichlorophenyl)triaz-2-ene-1-carbonitrile Potassium Salt
Shipping Condition Room Temperature
Molecular Formula C7 H3 Cl2 K N4
InChI Formula InChI=1S/C7H3Cl2N4.K/c8-6-2-1-5(3-7(6)9)12-13-11-4-10;/h1-3H;/q-1;+1
Solution Type Neat
SMILES Clc1ccc(cc1Cl)\N=N\N([K])C#N
IUPAC Name [cyano-[(E)-(3,4-dichlorophenyl)diazenyl]amino]potassium
Industry Type Pharmaceutical
Molecular Weight (g/mol) 253.13
Quantity 500 mg
Formula Weight 251.937
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.