missing translation for 'onlineSavingsMsg'
Learn More

3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%

Product Code. 10582781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10582781 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10582781 Supplier Thermo Scientific Acros Supplier No. 189160250
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25173-72-2 | C12H16O5 | 240.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25173-72-2
CAS Min % 98.0
CAS Max % 100.0
Melting Point 100.0°C to 105.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C12H16O5
Linear Formula (CH3O)3C6H2CH2CH2CO2H
MDL Number MFCD00002775
Quantity 25 g
Beilstein 10, III, 2120
Synonym 3-3,4,5-trimethoxyphenyl propanoic acid, 3-3,4,5-trimethoxyphenyl propionic acid, benzenepropanoic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyhydrocinnamic acid, 3,4,5-trimethoxydihydrocinnamic acid, 3,4,5-trimethoxyphenylpropionic acid, 3,4,5-trimethoxybenzenepropanoic acid, 3,4,5-trimethoxy-benzenepropanoic acid, acmc-1cazt, maybridge1_005958
InChI Key ZCYXGVJUZBKJAI-UHFFFAOYSA-N
SMILES COC1=CC(=CC(=C1OC)OC)CCC(=O)O
IUPAC Name 3-(3,4,5-trimethoxyphenyl)propanoic acid
Molecular Weight (g/mol) 240.25
PubChem CID 64860
ChEBI CHEBI:583580
Formula Weight 240.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(3, 4, 5-Trimethoxyphenyl)propionic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 3-(3,4,5-Trimethoxyphenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.