missing translation for 'onlineSavingsMsg'
Learn More

3,3',4',5-Tetrachlorosalicylanilide, 97%

Product Code. 10331691 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10331691

Brand: Thermo Scientific Acros 420230250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1154-59-2 | C13H7Cl4NO2 | 351.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1154-59-2
Molecular Formula C13H7Cl4NO2
Molecular Weight (g/mol) 351.01
MDL Number MFCD00041745
InChI Key SJQBHPJLLIJASD-UHFFFAOYSA-N
Synonym tcsa, 3,3',4',5-tetrachlorosalicylanilide, 3,5-dichloro-n-3,4-dichlorophenyl-2-hydroxybenzamide, 3,5,3',4'-tetrachlorosalicylanilide, irgasan bs-200, 3,5-dichlorosalicyl 3,4-dichloroanilide, caswell no. 833, 3,5-dichlorosalicyl-3,4-dichloroanilide, benzamide, 3,5-dichloro-n-3,4-dichlorophenyl-2-hydroxy, unii-hne676755i
PubChem CID 14385
ChEBI CHEBI:188648
IUPAC Name 3,5-dichloro-N-(3,4-dichlorophenyl)-2-hydroxybenzamide
SMILES C1=CC(=C(C=C1NC(=O)C2=CC(=CC(=C2O)Cl)Cl)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 162°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 9336
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alkaline aq. solution,soluble in many organic solvents
Formula Weight 351.01
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3,3',4',5-Tetrachlorosalicylanilide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3',4',5-Tetrachlorosalicylanilide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.