missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%

Product Code. 11467857
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467857 500 g 500g
11457857 100 g 100g
11389539 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467857 Supplier Thermo Scientific Alfa Aesar Supplier No. A16529.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2421-28-5 | C17H6O7 | 322.228 g/mol

BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.

Solubility
Soluble in DMF (0.1 g/mL). Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2421-28-5
Molecular Formula C17H6O7
Molecular Weight (g/mol) 322.228
MDL Number MFCD00005923
InChI Key VQVIHDPBMFABCQ-UHFFFAOYSA-N
Synonym 3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
PubChem CID 75498
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point 220°C to 226°C
Density 1.57
Boiling Point 320°C (5 mmHg)
Flash Point 324°C (615°F)
Quantity 500 g
Beilstein 761777
Sensitivity Moisture sensitive
Solubility Information Soluble in DMF (0.1g/mL). Reacts with water.
Formula Weight 322.23
Percent Purity ≥97%
Chemical Name or Material 3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Show More Show Less
compliance-icons
Product Identifier
  • 3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 3,3',4,4'-Benzophenonetetracarboxylic dianhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.