missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4',5,5'-Hexabromo-2,2'-diaminebiphenyl, TRC

Product Code. 30408749
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408749 1 mg 1mg
30391665 5 mg 5mg
30393729 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408749 Supplier Toronto Research Chemicals Supplier No. TRCH5899401MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3',4,4',5,5'-Hexabromo-2,2'-diaminebiphenyl
Analyte or Component Names 3,3',4,4',5,5'-Hexabromo-2,2'-diaminebiphenyl
Shipping Condition Room Temperature
Molecular Formula C12 H6 Br6 N2
Synonym 3,3',4,4',5,5'-Hexabromo-[1,1'-biphenyl]-2,2'-diamine
InChI Formula InChI=1S/C12H6Br6N2/c13-5-1-3(11(19)9(17)7(5)15)4-2-6(14)8(16)10(18)12(4)20/h1-2H,19-20H2
Solution Type Neat
SMILES Nc1c(Br)c(Br)c(Br)cc1c2cc(Br)c(Br)c(Br)c2N
IUPAC Name 6-(2-amino-3,4,5-tribromophenyl)-2,3,4-tribromoaniline
Industry Type Pharmaceutical
Molecular Weight (g/mol) 657.613
Quantity 1 mg
Formula Weight 651.563
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.