missing translation for 'onlineSavingsMsg'
Learn More

3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecanesulfonic Acid Sodium Salt, TRC

Product Code. 30011851
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011851 250 mg 250mg
17910425 25 mg 25mg
17900865 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011851 Supplier Toronto Research Chemicals Supplier No. TRCH245685250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108026-35-3
Molecular Formula C12 H4 F21 O3 S . Na
Molecular Weight (g/mol) 650.1796
Synonym 1H,1H,2H,2H-Perfluorododecane sulfonic acid sodium, 1-Dodecanesulfonic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-heneicosafluoro-, sodium salt (1:1)
IUPAC Name sodium;3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecane-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H5F21O3S.Na/c13-3(14,1-2-37(34,35)36)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)11(29,30)12(31,32)33;/h1-2H2,(H,34,35,36);/q;+1/p-1
Formula Weight 649.9444
Percent Purity >95%
Chemical Name or Material 1H,1H,2H,2H-Perfluorododecane sulfonic acid sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.