missing translation for 'onlineSavingsMsg'
Learn More

3,3,4,4,5,5,5-Heptafluoro-2-pentanol, 96%, Thermo Scientific™

Product Code. 11373746
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-14-4
Molecular Formula C5H5F7O
Molecular Weight (g/mol) 214.083
MDL Number MFCD00039619
InChI Key RBPHBIMHZSTIDT-UHFFFAOYSA-N
Synonym 3,3,4,4,5,5,5-heptafluoro-2-pentanol, acmc-20anhf, 3,3,4,4,5,5,5-heptafluoropentanol-2, 2-pentanol,3,3,4,4,5,5,5-heptafluoro, 2-pentanol, 3,3,4,4,5,5,5-heptafluoro, 3,3,4,4,5,5,5-heptakis fluoranyl pentan-2-ol
PubChem CID 78992
IUPAC Name 3,3,4,4,5,5,5-heptafluoropentan-2-ol
SMILES CC(C(C(C(F)(F)F)(F)F)(F)F)O

Specifications

Density 1.483
Boiling Point 101°C to 102°C
Refractive Index 1.312
Quantity 1 g
Beilstein 1705854
Formula Weight 214.08
Percent Purity 96%
Chemical Name or Material 3,3,4,4,5,5,5-Heptafluoro-2-pentanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'