missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3,3-Trifluoropropane-1-sulfonyl chloride, Maybridge
Specifications
Specifications
| CAS | 845866-80-0 |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C3H4ClF3O2S |
| Quantity | 1 g |
| Synonym | 3,3,3-trifluoro-1-propanesulfonyl chloride, 3,3,3-tris fluoranyl propane-1-sulfonyl chloride, chloro 3,3,3-trifluoropropyl sulfone, 3,3,3-trifluoropropylsulfonyl chloride, 3,3,3-trifluoropropanesulfonyl chloride, 3,3,3-trifluoropropylsulphonyl chloride, 3,3,3-trifluoropropane sulfonyl chloride, 3,3,3-trifluoropropane-1-sulphonyl chloride, 1-propanesulfonylchloride, 3,3,3-trifluoro, 3,3,3-trifluoro-1-propane sulfonyl chloride |
| InChI Key | OXKHPRFGCFJUSK-UHFFFAOYSA-N |
| Molecular Weight (g/mol) | 196.57 |
| PubChem CID | 2761177 |
| Formula Weight | 196.57 |
| Percent Purity | 97% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?