Learn More
3,3,3-Trifluorolactic acid, 96%, Thermo Scientific Chemicals
£33.75 - £131.00
Chemical Identifiers
CAS | 684-07-1 |
---|---|
Molecular Formula | C3H3F3O3 |
Molecular Weight (g/mol) | 144.049 |
MDL Number | MFCD00051802 |
InChI Key | BVKGUTLIPHZYCX-UHFFFAOYSA-N |
Synonym | 3,3,3-trifluorolactic acid, trifluorolactic acid, 2-hydroxy-3,3,3-trifluoropropanoic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy, 2-hydroxy-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-hydroxypropionic acid, acmc-20epmv, acmc-20dpo7, propanoic acid,3,3,3-trifluoro-2-hydroxy-, 2r, 3,3,3-trifluoro-dl-lactic acid |
PubChem CID | 244896 |
IUPAC Name | 3,3,3-trifluoro-2-hydroxypropanoic acid |
SMILES | C(C(=O)O)(C(F)(F)F)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,3,3-Trifluorolactic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, strong bases and water. Protect from humidity and water. Incompatible with oxidizing agents, bases.
Chemical Identifiers
684-07-1 | |
144.049 | |
BVKGUTLIPHZYCX-UHFFFAOYSA-N | |
244896 | |
C(C(=O)O)(C(F)(F)F)O |
C3H3F3O3 | |
MFCD00051802 | |
3,3,3-trifluorolactic acid, trifluorolactic acid, 2-hydroxy-3,3,3-trifluoropropanoic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy, 2-hydroxy-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-hydroxypropionic acid, acmc-20epmv, acmc-20dpo7, propanoic acid,3,3,3-trifluoro-2-hydroxy-, 2r, 3,3,3-trifluoro-dl-lactic acid | |
3,3,3-trifluoro-2-hydroxypropanoic acid |
Specifications
684-07-1 | |
157°C (123 mmHg) | |
MFCD00051802 | |
1907003 | |
3,3,3-trifluorolactic acid, trifluorolactic acid, 2-hydroxy-3,3,3-trifluoropropanoic acid, propanoic acid, 3,3,3-trifluoro-2-hydroxy, 2-hydroxy-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-hydroxypropionic acid, acmc-20epmv, acmc-20dpo7, propanoic acid,3,3,3-trifluoro-2-hydroxy-, 2r, 3,3,3-trifluoro-dl-lactic acid | |
BVKGUTLIPHZYCX-UHFFFAOYSA-N | |
3,3,3-trifluoro-2-hydroxypropanoic acid | |
244896 | |
96% |
66°C to 71°C | |
C3H3F3O3 | |
UN3261 | |
Hygroscopic | |
Soluble in water. | |
C(C(=O)O)(C(F)(F)F)O | |
144.049 | |
144.05 | |
3,3,3-Trifluorolactic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only