missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%

Product Code. 10739005 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10739005 5 g Glass bottle 5g
10491104 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10739005 Supplier Thermo Scientific Acros Supplier No. 397860050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 564-10-3 | C4H2F6O2 | 196.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 564-10-3
Molecular Formula C4H2F6O2
Molecular Weight (g/mol) 196.05
MDL Number MFCD00464165
InChI Key RAEAYTICAPHWJW-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid
PubChem CID 385606
IUPAC Name 3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
SMILES C(C(=O)O)(C(F)(F)F)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 50°C to 53°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (CF3)2CHCOOH
Quantity 5 g
Formula Weight 196.05
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3, 3-Trifluoro-2-(trifluoromethyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.