missing translation for 'onlineSavingsMsg'
Learn More

3,3,3-Trifluoro-2,2-dimethylpropionic acid, 97%

Product Code. 11301989
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301989 1 g 1g
15417545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301989 Supplier Thermo Scientific Alfa Aesar Supplier No. H26053.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 889940-13-0 | C5H7F3O2 | 156.104 g/mol

3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3,3-Trifluoro-2,2-dimethylpropionic acid is used in the synthesis of 2-aminobenzaldehyde oxime analogs which acts as dual inhibitors of neutrophil elastase and proteinase.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 889940-13-0
Molecular Formula C5H7F3O2
Molecular Weight (g/mol) 156.104
MDL Number MFCD08445819
InChI Key VMFSJVUPIXOCFO-UHFFFAOYSA-N
Synonym 3,3,3-trifluoro-2,2-dimethylpropionic acid, 2-trifluoromethyl-2-methylpropanoic acid, 3,3,3-trifluoro-2,2-dimethyl-propionic acid, 2-trifluoromethyl-isobutyric acid, 3,3,3-trifluoro-2,2-dimethyl-propanoic acid
PubChem CID 19867367
IUPAC Name 3,3,3-trifluoro-2,2-dimethylpropanoic acid
SMILES CC(C)(C(=O)O)C(F)(F)F

Specifications

Melting Point 66°C to 73°C
Boiling Point 76°C to 77°C (14 mmHg)
Quantity 1 g
UN Number UN3261
Formula Weight 156.12
Percent Purity 97%
Chemical Name or Material 3,3,3-Trifluoro-2,2-dimethylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.