missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3',3' '-(2,4,6-boroxintriyl)tris-Pyridine Hydrochloride (>70%), TRC
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
Specifications
Specifications
| Chemical Name or Material | 3,3',3''-(2,4,6-boroxintriyl)tris-Pyridine Hydrochloride |
| Molecular Formula | C15H12B3N3O3 •xHCl |
| Quantity | 100 mg |
| Synonym | 3,3',3''-(2,4,6-Boroxintriyl)tris[pyridine] Hydrochloride, 2,4,6-Tris(3-pyridinyl)cyclotriboroxane Hydrochloride, Tris(3-pyridyl)boroxin Hydrochloride |
| InChI Formula | InChI=1S/C15H12B3N3O3.ClH/c1-4-13(10-19-7-1)16-22-17(14-5-2-8-20-11-14)24-18(23-16)15-6-3-9-21-12-15;/h1-12H;1H |
| SMILES | B1(OB(OB(O1)C2=CC=CN=C2)C3=CC=CN=C3)C4=CN=CC=C4.Cl |
| IUPAC Name | 2,4,6-tri(pyridin-3-yl)-1,3,5,2,4,6-trioxatriborinane hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction