missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-((2-Hydroxypropane-1,3-diyl)bis(oxy))bis(4-chlorobenzoic acid) Diethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 3,3'-((2-Hydroxypropane-1,3-diyl)bis(oxy))bis(4-chlorobenzoic acid) Diethyl Ester |
| Analyte or Component Names | 3,3'-((2-Hydroxypropane-1,3-diyl)bis(oxy))bis(4-chlorobenzoic acid) Diethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H22 Cl2 O7 |
| InChI Formula | InChI=1S/C21H22Cl2O7/c1-3-27-20(25)13-5-7-16(22)18(9-13)29-11-15(24)12-30-19-10-14(6-8-17(19)23)21(26)28-4-2/h5-10,15,24H,3-4,11-12H2,1-2H3 |
| Solution Type | Neat |
| SMILES | CCOC(=O)c1ccc(Cl)c(OCC(O)COc2cc(ccc2Cl)C(=O)OCC)c1 |
| IUPAC Name | ethyl 4-chloro-3-[3-(2-chloro-5-ethoxycarbonylphenoxy)-2-hydroxypropoxy]benzoate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 457.301 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?