missing translation for 'onlineSavingsMsg'
Learn More

3,3'-((2-Hydroxypropane-1,3-diyl)bis(oxy))bis(4-chlorobenzoic acid) Diethyl Ester, TRC

Product Code. 30403412
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403412 50 mg 50mg
30399880 25 mg 25mg
30403776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403412 Supplier Toronto Research Chemicals Supplier No. TRCH95272050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 3,3'-((2-Hydroxypropane-1,3-diyl)bis(oxy))bis(4-chlorobenzoic acid) Diethyl Ester
Analyte or Component Names 3,3'-((2-Hydroxypropane-1,3-diyl)bis(oxy))bis(4-chlorobenzoic acid) Diethyl Ester
Shipping Condition Room Temperature
Molecular Formula C21 H22 Cl2 O7
InChI Formula InChI=1S/C21H22Cl2O7/c1-3-27-20(25)13-5-7-16(22)18(9-13)29-11-15(24)12-30-19-10-14(6-8-17(19)23)21(26)28-4-2/h5-10,15,24H,3-4,11-12H2,1-2H3
Solution Type Neat
SMILES CCOC(=O)c1ccc(Cl)c(OCC(O)COc2cc(ccc2Cl)C(=O)OCC)c1
IUPAC Name ethyl 4-chloro-3-[3-(2-chloro-5-ethoxycarbonylphenoxy)-2-hydroxypropoxy]benzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 457.301
Quantity 50 mg
Formula Weight 456.074
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.