missing translation for 'onlineSavingsMsg'
Learn More

3-(25-Hydroxy Vitamin D3)propylamino-PEG12-Biotin, TRC

Product Code. 30401223
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401223 10 mg 10mg
30397964 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30401223 Supplier Toronto Research Chemicals Supplier No. TRCH99590010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(25-Hydroxy Vitamin D3)propylamino-PEG12-Biotin
Analyte or Component Names 3-(25-Hydroxy Vitamin D3)propylamino-PEG12-Biotin
Shipping Condition Dry ice
Molecular Formula C67 H118 N4 O17 S
InChI Formula InChI=1S/C67H118N4O17S/c1-53-15-18-57(51-56(53)17-16-55-12-9-23-67(5)58(19-20-59(55)67)54(2)11-8-22-66(3,4)75)88-26-10-24-68-63(73)21-27-76-29-31-78-33-35-80-37-39-82-41-43-84-45-47-86-49-50-87-48-46-85-44-42-83-40-38-81-36-34-79-32-30-77-28-25-69-62(72)14-7-6-13-61-64-60(52-89-61)70-65(74)71-64/h16-17,54,57-61,64,75H,1,6-15,18-52H2,2-5H3,(H,68,73)(H,69,72)(H2,70,71,74)/b55-16+,56-17-/t54-,57+,58-,59+,60+,61+,64+,67-/m1/s1
Solution Type Neat
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@H](CCC3=C)OCCCNC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)CCCC[C@@H]4SC[C@@H]5NC(=O)N[C@H]45)\CCC[C@]12C
IUPAC Name N-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[3-[3-[(1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-5-hydroxy-1,5-dimethyl-hexyl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylene-cyclohexoxy]propylamino]-3-oxo-propoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethyl]-5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanamide
Industry Type Forensics
Molecular Weight (g/mol) 1283.74
Quantity 10 mg
Formula Weight 1282.8213
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.