missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)propanoic acid, 98%

Product Code. 10182481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182481 5 g Glass bottle 5g
10603302 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182481 Supplier Thermo Scientific Acros Supplier No. 330110050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5928-51-8 | C7H8O2S | 156.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5928-51-8
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.2
MDL Number MFCD00047093
InChI Key MJPVYTKZYZPIQA-UHFFFAOYSA-N
Synonym 3-2-thienyl propanoic acid, 3-2-thienyl propionic acid, 2-thiophenepropanoic acid, 3-thiophen-2-yl propanoic acid, 2-thiophenepropionic acid, maybridge3_005592, 3-2-thienyl propionic pound inverted question markacid, 3-thiophen-2-yl-propionic acid, thiolpropionic acid, pubchem23928
PubChem CID 703169
IUPAC Name 3-thiophen-2-ylpropanoic acid
SMILES C1=CSC(=C1)CCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 45°C to 49°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol, acetone
Formula Weight 156.2
Percent Purity 98%
Physical Form Crystals or Needles
Chemical Name or Material 3-(2-Thienyl)propanoic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.